Phosphorus(III) bromide 99%
|
Danger
|
Synonyms:
Phosphorus tribromide
AC16947-1000
103526-266EA
64.8
USD
103526-266
Phosphorus(III) bromide 99%
Phosphorus (III) bromide
Formula:
PBr₃ MW: 270.69 g/mol Boiling Pt: 175 °C (760 mmHg) Melting Pt: –40 °C Density: 2.88 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011436 CAS Number: 7789-60-8 EINECS: 232-178-2 UN: 1808 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...