Peanut oil
AC41685-5000
AC41685-0025
103467-396EA
58.8
USD
103467-396
103467-394
Peanut oil
Peanut oil
Density:
0.915 g/cm³ Flash Pt: 283 °C Storage Temperature: Ambient |
MDL Number:
MFCD00131805 CAS Number: 8002-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...