Patent blue V sodium salt

Synonyms: Food Blue 5 sodium saltBis[hydrogen [4-[4-(diethylamino)-5'-hydroxy-2',4'-disulfonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium], sodium saltC.I. 42051Bis[hydrogen [4-[4-(diethylamino)-5'-hydroxy-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium], sodium saltAcid blue 3 sodium salt

AC33933-1000 AC33933-0050 AC33933-0250
103518-720EA 35.4 USD
103518-720 103518-722 103518-718
Patent blue V sodium salt
Patent blue V sodium salt
Formula: C₂₇H₃₁N₂NaO₇S₂
MW: 582.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09608138
CAS Number: 20262-76-4
EINECS: 222-573-8

Order Now

Specification Test Results

Appearance Dark blue-green Crystalline powder
Infrared spectrum Conforms
Dye content ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR