Paraffin, liquid, pure

Warning

Synonyms: Wax ParaffinParaffin wax

AC17140-0010 AC17140-0025 AC17140-2500
103467-388EA 65.4 USD
103467-388 103467-392 103467-390
Paraffin, liquid, pure
Paraffin
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: –24 °C
Density: 0.88 g/cm³
Flash Pt: 218 °C
Storage Temperature: Ambient
CAS Number: 8002-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR