o-Xylene, extra dry 99%, AcroSeal™
|
Danger
|
Synonyms:
1,2-Dimethylbenzene
AC44302-0010
103524-500EA
123.46
USD
103524-500
o-Xylene, extra dry 99%, AcroSeal™
o-Xylene
Formula:
C₈H₁₀ MW: 106.17 g/mol Boiling Pt: 143…145 °C (760 mmHg) Melting Pt: –25 °C Density: 0.878 g/cm³ Flash Pt: 31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008519 CAS Number: 95-47-6 UN: 1307 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.005 % |
GC | ≥98.5 % |
Residue after evaporation | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...