4-Oxazolecarboxylic acid 97%

Warning

Synonyms: Oxazole-4-carboxylic acid

AC43011-0010 AC43011-0050
103515-152EA 340 USD
103515-152 103515-150
4-Oxazolecarboxylic acid 97%
4-Oxazolecarboxylic acid
Formula: C₄H₃NO₃
MW: 113.0715 g/mol
Melting Pt: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD06797151
CAS Number: 23012-13-7

Order Now

Specification Test Results

Appearance Light orange to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR