Oleanolic acid 97%

Warning

AC45612-0050
103517-678EA 67.1 USD
103517-678
Oleanolic acid 97%
Oleanolic acid
Formula: C₃₀H₄₈O₃
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064914
CAS Number: 508-02-1

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR