O-Benzyl-L-serine 99%

Synonyms: H-Ser(Bzl)-OH

AC44093-0250
103516-164EA 340 USD
103516-164
O-Benzyl-L-serine 99%
O-Benzyl-L-serine
Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Melting Pt: 210…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00065937
CAS Number: 4726-96-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 %
HPLC ≥98.5 %
TLC ≥98.5 %
Specific optical rotation +20.0° to +25.0° (20 °C, 589 nm) (c=1 in 8:2 AcOH/1M HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR