Benzyl N-(2-aminoethyl)carbamate hydrochloride 95%

Warning

Synonyms: N-1-Z-1,2-diaminoethane hydrochlorideN-(2-Aminoethyl)carbamic acid benzyl ester hydrochlorideN-Carbobenzoxy-1,2-diaminoethane hydrochlorideN-Z-Ethylenediamine hydrochlorideN-Cbz-1,2-diaminoethane hydrochloride

AC43981-0010
103516-042EA 48.4 USD
103516-042
Benzyl N-(2-aminoethyl)carbamate hydrochloride 95%
N-Carbobenzoxy-1,2-diaminoethane hydrochloride
Formula: C₁₀H₁₄N₂O₂
MW: 230.69 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00270150
CAS Number: 18807-71-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR