N-Phenylrhodanine 97+%
|
Warning
|
Synonyms:
3-Phenyl-2-thioxo-1,3-thiazolidin-4-one
AC39139-0050
AC39139-0010
103514-100EA
31.4
USD
103514-100
103514-102
N-Phenylrhodanine 97+%
N-Phenylrhodanine
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 190 to 194 °C |
HPLC | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...