9-Phenylcarbazole 97%

Danger

Synonyms: N-Phenylcarbazole

AC13040-0050 AC13040-0250 AC13040-1000
103466-196EA 345 USD
103466-196 103466-194 103466-192
9-Phenylcarbazole 97%
9-Phenylcarbazole
Formula: C₁₈H₁₃N
MW: 243.30 g/mol
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004965
CAS Number: 1150-62-5

Order Now

Specification Test Results

Appearance Off-white to brown Powder
Infrared spectrum Conforms
Melting point 93 to 98 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR