1-Phenacylpyridinium bromide 97%
|
Warning
|
Synonyms:
1-(2-Oxo-2-phenylethyl)pyridin-1-ium bromide
, N-Phenacylpyridinium bromide
, N-(Benzoylmethyl)pyridinium bromide
AC45012-0100
AC45012-0500
103517-176EA
22.7
USD
103517-176
103517-178
1-Phenacylpyridinium bromide 97%
1-Phenacylpyridinium bromide
Formula:
C₁₃H₁₂BrNO MW: 278.15 g/mol Melting Pt: 193…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031995 CAS Number: 16883-69-5 |
Specification Test Results
Appearance | Off-white to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 196 to 200 °C |
Titration Argentometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...