n-Pentane for analysis

Danger

Synonyms: Pentane

AC38390-0100 AC38390-0025 AC38390-0010
103528-206EA 1014.23 USD
103528-206 103528-204 103528-202
n-Pentane for analysis
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: –130 °C
Density: 0.626 g/cm³
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
https://us.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.01 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.0001 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.1 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.1 ppm
Total phosphorus (P) ≤0.5 ppm
Total silicon (Si) ≤0.02 ppm
Total sulfur (S) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR