Sodium octadecyl sulfate 98%
|
Warning
|
Synonyms:
Octadecyl sulphuric acid salt
, Sodium Stearyl sulphate
, Sodium Stearyl sulfate
, Sodium n-octadecylsulphate
, n-Octadecyl sulphate, sodium salt
, Octadecyl sulfuric acid salt
, n-Octadecyl sulfate, sodium salt
, Sodium n-octadecylsulfate
AC38489-0010
103466-946EA
62.6
USD
103466-946
Sodium octadecyl sulfate 98%
Sodium octadecyl sulphate
Formula:
C₁₈H₃₇NaO₄S MW: 372.55 g/mol Melting Pt: 211…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007469 CAS Number: 1120-04-3 |
Specification Test Results
Appearance | White to light yellow to light beige Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤7 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...