N-Methyl-N-prop-2-ynylamine 98+%

Danger

Synonyms: N-Methylpropargylamine

AC16554-0010 AC16554-0050
103526-146EA 169 USD
103526-146 103526-144
N-Methyl-N-prop-2-ynylamine 98+%
N-Methyl-N-prop-2-ynylamine
Formula: C₄H₇N
MW: 69.11 g/mol
Boiling Pt: 82…84 °C (760 mmHg)
Density: 0.83 g/cm³
Flash Pt: 1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008573
CAS Number: 35161-71-8
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR