1-Methylindole-3-boronic acid pinacol ester 97%
|
Warning
|
Synonyms:
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
, N-Methylindole-3-boronic acid pinacol ester
AC44101-0050
103466-170EA
832
USD
103466-170
1-Methylindole-3-boronic acid pinacol ester 97%
1-Methylindole-3-boronic acid pinacol ester
Formula:
C₁₅H₂₀BNO₂ MW: 257.1358 g/mol Melting Pt: 106…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD11894338 CAS Number: 683229-61-0 |
Specification Test Results
Appearance | White to light orange to light brown Solid |
Infrared spectrum | Conforms |
Melting point | 105 to 113 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...