N-Methoxy-N-methylbenzamide 98%

Warning

AC43656-0050 AC43656-0250
103466-158EA 250.4 USD
103466-158 103466-156
N-Methoxy-N-methylbenzamide 98%
N-Methoxy-N-methylbenzamide
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 85 °C (1 mmHg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075320
CAS Number: 6919-61-5

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR