N-Lauroylsarcosine sodium salt 95%

Danger

Synonyms: SarkosylSodium lauroyl sarcosineSodium N-lauroylsarcosinate

AC43437-0250 AC43437-1000 AC43437-0010
103524-404EA 405 USD
103524-404 103524-406 103524-408
N-Lauroylsarcosine sodium salt 95%
N-Lauroylsarcosine sodium salt
Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MW: 293.38 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2811
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/52/52/2355252.jpg

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Water ≤7 %
Solubility (10 % in water) Almost clear colorless to light yellow
pH 7.0 to 9.0
Titration with HClO4 94 to 106 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR