N-Hydroxysulfosuccinimide sodium salt (Sulfo-NHS) 95%

Warning

Synonyms: N-Hydroxysulfosuccinimide sodium saltsulfo-N-HydroxysuccinimideHydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium saltN-Hydroxy sulfosuccinimide sodium saltSulfo NHSN-Hydroxysulphosuccinimide sodium saltHydroxy-2,5-dioxopyrrolidine-3-sulfonic acid sodium saltN-Hydroxy sulphosuccinimide sodium saltSulpho-N-Hydroxysuccinimide

AC43865-0010
103466-942EA 313 USD
103466-942
N-Hydroxysulfosuccinimide sodium salt (Sulfo-NHS) 95%
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt)
Formula: C₄H₄NNaO₆S
MW: 217.13 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Sodium (Na) 9.95 to 11.22 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR