n-Hexyllithium 33% (w/w) 2.5 N in hexane, AcroSeal™
|
Danger
|
Synonyms:
lithium hexane
AC30165-1000
AC30165-8000
103527-440EA
246.62
USD
103527-440
103527-438
n-Hexyllithium 33% (w/w) 2.5 N in hexane, AcroSeal™
n-Hexyllithium
Formula:
C₆H₁₃Li MW: 92.11 g/mol Density: 0.69 g/cm³ Flash Pt: –26 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00191446 CAS Number: 21369-64-2 UN: 3394 ADR: 4.2, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...