n-Hexane contains appr. 95% n-Hexane for analysis

Danger

Synonyms: HexanenHexaneHexyl hydride

AC38380-0010 AC38380-0250 AC38380-0050 AC38380-0025
103528-194EA 251 USD
103528-194 103528-192 103528-190 103528-196
n-Hexane contains appr. 95% n-Hexane for analysis
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.02 % (K.F.)
GC ≥95.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.00008 meq/g
Substance darkened by H2SO4 passes test
Calcium (Ca) ≤0.2 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Hydrocarbons ≤0.1 % Aromatic hydrocarbons
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤10 ppm
Bromine number ≤0.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR