Hexadecyl palmitate 95%

Synonyms: Palmityl palmitateCetyl Palmitaten-Hexadecyl palmitatePalmitic Acid Hexadecyl EsterPalmitic acid palmityl ester

AC42822-2500 AC42822-0010
103515-026EA 30.5 USD
103515-026 103515-024
Hexadecyl palmitate 95%
Hexadecyl palmitate
Formula: C₃₂H₆₄O₂
MW: 480.86 g/mol
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00053739
CAS Number: 540-10-3

Order Now

Specification Test Results

Appearance White to off-white Waxy flakes or platelets
Infrared spectrum Conforms
GC ≥94 %
NMR (31P-NMR) ≥99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR