Boc-Lys(Fmoc)-OH 99%

Warning

Synonyms: N-epsilon-FMOC-N-alpha-BOC-L-Lysine

AC43908-0050
103466-132EA 145 USD
103466-132
Boc-Lys(Fmoc)-OH 99%
Boc-Lys(Fmoc)-OH
Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Melting Pt: 135…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062032
CAS Number: 84624-27-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +2.3° to +4.3° (20 °C, 589 nm) (c=1 in Ethyl acetate)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR