N-Benzylideneaniline 99%

Warning

AC43778-0250 AC43778-0050
103513-206EA 51 USD
103513-206 103513-208
N-Benzylideneaniline 99%
N-Benzylideneaniline
Formula: C₁₃H₁₁N
MW: 181.24 g/mol
Boiling Pt: 300 °C
Melting Pt: 52…54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003027
CAS Number: 538-51-2

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 55 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR