N-α-Boc-N-ε-(2-chloro-Z)-L-lysine 98%
Synonyms:
Boc-Lys(2-chloro-Z)-OH
, Boc-Lys(2-Cl-Z)-OH
AC44087-0050
103516-150EA
42.8
USD
103516-150
N-α-Boc-N-ε-(2-chloro-Z)-L-lysine 98%
N-alpha-Boc-N-epsilon-(2-chloro-Z)-L-lysine
Formula:
C₁₉H₂₇ClN₂O₆ MW: 414.883 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00038386 CAS Number: 54613-99-9 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤1.00 % |
HPLC | ≥97.5 % |
TLC | ≥97.5 % |
Specific optical rotation | -10.8° to -11.8° (20 °C, 589 nm) (c=2 in DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...