N-α-Boc-N-β-xanthenyl-L-asparagine 96%

Synonyms: Boc-Asn(Xan)-OH

AC44084-0050 AC44084-0010
103466-012EA 144.4 USD
103466-012 103466-010
N-α-Boc-N-β-xanthenyl-L-asparagine 96%
N-alpha-Boc-N-beta-xanthenyl-L-asparagine
Formula: C₂₂H₂₄N₂O₆
MW: 412.442 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00066157
CAS Number: 65420-40-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 %
HPLC ≥95.0 %
TLC ≥95 %
Appearance of solution (0.5mmol/ml DMF) clear, colourless
Specific optical rotation +9.0° to +11.0° (20 °C, 589 nm) (c=1 in MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR