N(α)-Boc-L-histidine 99+%

Synonyms: Boc-His-OH

AC27559-0010
103512-012EA 27.5 USD
103512-012
N(α)-Boc-L-histidine 99+%
N(α)-Boc-L-histidine
Formula: C₁₁H₁₇N₃O₄
MW: 255.3 g/mol
Melting Pt: 196…199 °C
Storage Temperature: Ambient
EINECS: 241-768-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR