(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 98%

Warning

Synonyms: Propanamide, 2-[(5-fluoro-2,4-dinitrophenyl)amino]-, (2S)-N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide1-Fluoro-2,4-dinitrophenyl-5-L-alanine amide

AC43934-1000
103465-998EA 253 USD
103465-998
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 98%
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
Formula: C₉H₉FN₄O₅
MW: 272.192 g/mol
Melting Pt: 227.7…229.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042049
CAS Number: 95713-52-3

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +54.0° to +58.0° (20 °C, 589 nm) (c=1, acetone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR