N-Succinimidyl Acrylate 99%

Warning

Synonyms: N-AcryloyloxysuccinimideAcrylic Acid N-Succinimidyl EsterN-Acryloxysuccinimide2,5-Dioxopyrrolidin-1-yl acrylate

AC40030-0010 AC40030-0250
103518-822EA 909 USD
103518-822 103518-818
N-Succinimidyl Acrylate 99%
N-Succinimidyl Acrylate
Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 69 °C
Storage Temperature: Freezer
MDL Number: MFCD00078261
CAS Number: 38862-24-7

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR