N-Ethyldiisopropylamine 99.5+%, AcroSeal™

Danger

Synonyms: N.N’-Diisopropyl ethylamineN,N-DiisopropylethylamineDIPEAEthyldiisopropylamineHünig’s base

AC45959-0010 AC45959-1000
103529-068EA 98.9 USD
103529-068 103529-066
N-Ethyldiisopropylamine 99.5+%, AcroSeal™
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 127 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.756 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
UN: 3384
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4150 (20 °C, nm)
Water ≤0.3 %
GC ≥99.50 %
Color scale ≤50 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR