Diisopropylcarbodiimide 99%, AcroSeal™

Danger

Synonyms: N,N'-methanetetraylbis(1-methylethylamine)1,3-DiisopropylcarbodiimideN,N-DiisopropylcarbodiimideN,N'-DiisopropylcarbodiimideN.N’-DiisopropylcarbodiimideDIC

AC44618-1000
103524-556EA 185 USD
103524-556
Diisopropylcarbodiimide 99%, AcroSeal™
Diisopropylcarbodiimide
Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 145…148 °C (760 mmHg)
Density: 0.81 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00065689
CAS Number: 693-13-0
UN: 3384
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4350 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR