N,N-Diisopropylcarbamoyl chloride 98%

Danger

Synonyms: Diisopropylcarbamoyl Chloride

AC31452-0050 AC31452-0250
103527-524EA 51.5 USD
103527-524 103527-526
N,N-Diisopropylcarbamoyl chloride 98%
N,N-Diisopropylcarbamoyl chloride
Formula: C₇H₁₄ClNO
MW: 163.6452 g/mol
Boiling Pt: 90…93 °C (15 mmHg)
Melting Pt: 58.1…59.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015013
EINECS: 242-743-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR