N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 99%

Synonyms: N,N,N,N-Tetrakis(2-hydroxyethyl)ethylenediamine

AC42048-0010 AC42048-5000 AC42048-1000
103512-524EA 35.2 USD
103512-524 103512-526 103514-902
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
Formula: C₁₀H₂₄N₂O₄
MW: 236.3086 g/mol
Density: 1.13 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
EINECS: 205-396-0

Order Now

Specification Test Results

Appearance Clear colorless to amber Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4990 to 1.5070 (20 °C, 589 nm)
pH ≥10 (25 % solution)
Titration with HClO4 ≥98.5 %
Arsenic (As) ≤2 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤400 ppm
Lead (Pb) ≤5 ppm
Mercury (Hg) ≤1 ppm
Zinc (Zn) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.2 % (in HCl)
Loss on ignition ≤8 % (800 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR