N-(2-Chloroethyl)dibenzylamine hydrochloride 98%

Warning

Synonyms: N,N-Dibenzyl-2-chloroethylamine HydrochlorideDibenamine HydrochlorideN,N-Dibenzylaminoethyl Chloride Hydrochloride

AC43630-0050 AC43630-0250
103515-794EA 237.1 USD
103515-794 103515-792
N-(2-Chloroethyl)dibenzylamine hydrochloride 98%
N-(2-Chloroethyl)dibenzylamine hydrochloride
Formula: C₁₆H₁₈ClN
MW: 296.24 g/mol
Melting Pt: 188…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00012518
CAS Number: 55-43-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 188 to 195 °C
Elemental Analysis 63.2 to 66.5 % C
Elemental Analysis 4.5 to 4.9 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR