Montelukast sodium 98%

Danger

Synonyms: Sodium (R,E)-2-(1-(((1-(3-(2-(7-chloroquinolin-2-yl)vinyl)phenyl)-3-(2-(2-hydroxypropan-2-yl)phenyl)propyl)thio)methyl)cyclopropyl)acetate

AC45644-0010
103517-496EA 179 USD
103517-496
Montelukast sodium 98%
Montelukast sodium
Formula: C₃₅H₃₆ClNO₃S
MW: 608.17 g/mol
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00931431
CAS Number: 151767-02-1

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms
Water ≤2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR