Methyl linoleate 99%

Synonyms: (9Z,12Z)-Methyl octadeca-9,12-dienoateLinoleic acid methyl estercis,cis-9,12-Octadecadienoic acid methyl ester9Z,12Z-Octadecadienoic acid methyl ester

AC42813-0250 AC42813-1000 AC42813-0050
103465-886EA 68.8 USD
103465-886 103465-888 103465-890
Methyl linoleate 99%
Methyl linoleate
Formula: C₁₉H₃₄O₂
MW: 294.48 g/mol
Boiling Pt: 212 °C (16 mmHg)
Melting Pt: –35 °C
Density: 0.89 g/cm³
Flash Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009534
CAS Number: 112-63-0
EINECS: 203-993-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation -85° to -95° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR