Methyl indole-6-carboxylate 97%

Warning

Synonyms: Methyl 1H-indole-6-carboxylate

AC39793-0010 AC39793-0050
103514-488EA 33.8 USD
103514-488 103514-490
Methyl indole-6-carboxylate 97%
Methyl indole-6-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 76…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00211063
CAS Number: 50820-65-0

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 79 to 84 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR