Methyl phenylglyoxylate 99%

Synonyms: Methyl benzoylformateMethyl 2-oxo-2-phenylacetate

AC12637-0050 AC12637-0250
103510-114EA 42.9 USD
103510-114 103510-116
Methyl phenylglyoxylate 99%
Methyl phenylglyoxylate
Formula: C₉H₈O₃
MW: 164.1600 g/mol
Boiling Pt: 246…248 °C (760 mmHg)
Melting Pt: 16 °C
Density: 1.15 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008443
EINECS: 239-263-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5280 (20 °C, 589 nm)
GC ≥98.5 %
Substance darkened by H2SO4 Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR