Methyl-6-amino-5-bromonicotinate 98%

Warning

Synonyms: Methyl 6-amino-5-bromonicotinate

AC43794-0050 AC43794-0010
103513-222EA 321 USD
103513-222 103513-220
Methyl-6-amino-5-bromonicotinate 98%
Methyl 6-amino-5-bromonicotinate
Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 173.5…175 °C
Storage Temperature: Ambient
MDL Number: MFCD06659561
CAS Number: 180340-70-9

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
Melting point 172 to 176 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR