Methyl-5-hydroxynicotinate 97+%

Warning

Synonyms: Methyl 5-hydroxynicotinateMethyl 5-hydroxy-3-pyridinecarboxylate

AC39335-0010 AC39335-0050 AC39335-2500
103514-364EA 48.5 USD
103514-364 103514-360 103514-362
Methyl-5-hydroxynicotinate 97+%
Methyl 5-hydroxynicotinate
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 195…197 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191521
CAS Number: 30766-22-4

Order Now

Specification Test Results

Appearance Light beige Powder and/or lumps
Infrared spectrum Conforms
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR