Methyl-5-hexynoate 95%

Warning

Synonyms: Methyl 5-hexynoate

AC38088-0050 AC38088-0250
103513-124EA 385 USD
103513-124 103513-122
Methyl-5-hexynoate 95%
Methyl-5-hexynoate
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 60…62 °C (10 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00671366
CAS Number: 77758-51-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR