Methyl-3,3-dimethoxypropionate 99%

Warning

Synonyms: Methyl 3,3-dimethoxypropanoate

AC26434-0250 AC26434-0050
103511-448EA 75.4 USD
103511-448 103511-446
Methyl-3,3-dimethoxypropionate 99%
Methyl 3,3-dimethoxypropionate
Formula: C₆H₁₂O₄
MW: 148.16 g/mol
Boiling Pt: 166 °C (760 mmHg)
Density: 1.06 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
EINECS: 231-055-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR