Methyl-2-methylnicotinate 97%

Warning

Synonyms: Methyl 2-methylnicotinatemethyl 2-methylpyridine-3-carboxylate

AC44925-0250
103517-082EA 187 USD
103517-082
Methyl-2-methylnicotinate 97%
Methyl 2-methylnicotinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 102…104 °C (10 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00051961
CAS Number: 65719-09-7

Order Now

Specification Test Results

Appearance Colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5145 to 1.5185 (20 °C, 589 nm)
Water ≤1.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR