Methyl-2,4-dichlorobenzoate 97%

Warning

Synonyms: Methyl 2,4-dichlorobenzoate

AC43005-0050 AC43005-0250
103515-134EA 112 USD
103515-134 103515-132
Methyl-2,4-dichlorobenzoate 97%
Methyl 2,4-dichlorobenzoate
Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
Boiling Pt: 132 °C (15 mmHg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00085117
CAS Number: 35112-28-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR