Methyl (R)-(-)-3-hydroxyisobutyrate 98%

Synonyms: (R)-Methyl 3-hydroxy-2-methylpropanoateMethyl (R)-(-)-beta-hydroxyisobutyrate

AC43618-0010
103515-768EA 45.55 USD
103515-768
Methyl (R)-(-)-3-hydroxyisobutyrate 98%
Methyl (R)-(-)-3-hydroxyisobutyrate
Formula: C₅H₁₀O₃
MW: 118.1311 g/mol
Boiling Pt: 60 °C (4Torr)
Density: 1.07 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00063450
CAS Number: 72657-23-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation -23° to -29° (20 °C, 589 nm) (c=4, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR