Methoxyacetonitrile 98%

Warning

AC12566-0250 AC12566-0050 AC12566-1000
103525-262EA 70.4 USD
103525-262 103525-264 103525-260
Methoxyacetonitrile 98%
Methoxyacetonitrile
Formula: C₃H₅NO
Boiling Pt: 118…119 °C (731 mmHg)
Density: 0.95 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00001892
CAS Number: 1738-36-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3800 to 1.3830 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR