Methanol-[D₄] (99.8% D) + 0.03% (v/v) TMS, AcroSeal™ for NMR spectroscopy

Danger

Synonyms: Carbinol-D4Methyl alcohol-D4Methanol-[²H₄]Methyl-D3 alcohol-DWood alcohol-D4Methanol-d4Hydroxy methane-D4Tetradeuteromethanol

AC32535-1000
103527-628EA 733 USD
103527-628
Methanol-[D₄] (99.8% D) + 0.03% (v/v) TMS, AcroSeal™ for NMR spectroscopy
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65 °C (760 mmHg)
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR