Methacrolein 90% stabilized

Danger

Synonyms: Acrolein

AC29283-1000 AC29283-5000 AC29283-0500 AC29283-0100
103527-340EA 423 USD
103527-340 103527-342 103527-336 103527-338
Methacrolein 90% stabilized
Methacrolein
Formula: C₄H₆O
Boiling Pt: 69 °C (760 mmHg)
Melting Pt: –81 °C
Density: 0.847 g/cm³
Flash Pt: –15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006974
CAS Number: 78-85-3
EINECS: 201-150-1
UN: 2396
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR