Methacrolein 90% stabilized, AcroSeal™
|
Danger
|
Synonyms:
Acrolein
AC19650-1000
103526-486EA
421
USD
103526-486
Methacrolein 90% stabilized, AcroSeal™
Methacrolein
Formula:
C₄H₆O Boiling Pt: 69 °C (760 mmHg) Melting Pt: –81 °C Density: 0.847 g/cm³ Flash Pt: –15 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006974 CAS Number: 78-85-3 UN: 2396 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Stabilizer | 0.1 to 0.5 % Hydroquinone |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...