Lopinavir 98%

Synonyms: (αS)-N-[(1S,3S,4S)-4-[[(2,6-Dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-α-(1-methylethyl)-2-oxo-1(2H)-(S)-N-((2R,4S,5S)-5-(2-(2,6-Dimethylphenoxy)acetamido)-4-hydroxy-1,6-diphenylhexn-2-yl)-3-methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanamide

AC45675-0050 AC45675-0010
103518-412EA 519 USD
103518-412 103518-410
Lopinavir 98%
Lopinavir
Formula: C₃₇H₄₈N₄O₅
MW: 628.82 g/mol
Melting Pt: 124…127 °C
Storage Temperature: Ambient
MDL Number: MFCD22373646
CAS Number: 192725-17-0

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Water ≤4 %
HPLC ≥97.5 %
Residue after ignition ≤0.2 %
Specific optical rotation -22° to -27° (20 °C, 589 nm) (c=0.8, methanol)
Specific optical rotation (On anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR